4-Nitro-3-(trifluoromethyl)benzenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Nitro-3-(trifluoromethyl)benzene-1-sulphonyl chloride
, 4-Nitro-3-(trifluoromethyl)benzene-1-sulfonyl chloride
L19721-06
L19721-03
L19721-06
L19721-03
AAAL19721-06EA
162.99
USD
AAAL19721-06
CAAAAL19721-03
CAAAAL19721-06
AAAL19721-03
4-Nitro-3-(trifluoromethyl)benzenesulfonyl chloride 97%
4-Nitro-3-(trifluoromethyl)benzenesulphonyl chloride
Formula:
C₇H₃ClF₃NO₄S MW: 289.62 g/mol Melting Pt: 63…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094387 CAS Number: 39234-83-8 EINECS: 000-000-0 UN: 3261 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...