(S)-(-)-1-Benzyl-3-acetamidopyrrolidine 98%, ee 99%

L19689-06 L19689-03 L19689-03 L19689-06
AAAL19689-03EA 108.63 USD
CAAAAL19689-06 AAAL19689-03 AAAL19689-06 CAAAAL19689-03
(S)-(-)-1-Benzyl-3-acetamidopyrrolidine 98%, ee 99%
(S)-(-)-1-Benzyl-3-acetamidopyrrolidine
Formula: C₁₃H₁₈N₂O
Melting Pt: 44…46 °C
Storage Temperature: Ambient
MDL Number: MFCD03788746
CAS Number: 114636-30-5

Order Now

Specification Test Results

Optical rotation -38° (c=1 in methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR