(E)-1-Hexene-1,2-diboronic acid bis(pinacol)ester 98%

L19650-06 L19650-06 L19650-03 L19650-03
AAAL19650-06EA 261.96 USD
AAAL19650-06 CAAAAL19650-06 AAAL19650-03 CAAAAL19650-03
(E)-1-Hexene-1,2-diboronic acid bis(pinacol)ester 98%
(E)-1-Hexene-1,2-diboronic acid bis(pinacol) ester
Formula: C₁₈H₃₄B₂O₄
Boiling Pt: 102…104 °C/0.08 mm
Density: 0.954
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03093898
CAS Number: 185427-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR