2-(9-Carbazolyl)ethylboronic acid diethanolamine ester 98%
L19575-MC
L19575-MC
AAAL19575-MCEA
60.51
USD
CAAAAL19575-MC
AAAL19575-MC
2-(9-Carbazolyl)ethylboronic acid diethanolamine ester 98%
2-(9-Carbazolyl)ethylboronic acid diethanolamine ester
Formula:
C₁₈H₂₁BN₂O₂ Melting Pt: 244…248 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788733 CAS Number: 501014-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...