4,4-Difluoropiperidine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19555-03
L19555-MD
L19555-03
AAAL19555-03EA
130.99
USD
CAAAAL19555-MD
CAAAAL19555-03
AAAL19555-03
4,4-Difluoropiperidine hydrochloride 98%
4,4-Difluoropiperidine hydrochloride
Formula:
C₅H₁₀ClF₂N MW: 157.59 g/mol Melting Pt: ca 172 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095381 CAS Number: 144230-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...