3-Bromo-4-nitropyridine-N-oxide 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromo-4-nitropyridine 1-oxide

L19524-06 L19524-14 L19524-03 L19524-06 L19524-03
AAAL19524-03EA 42.22 USD
CAAAAL19524-14 CAAAAL19524-03 AAAL19524-03 AAAL19524-06 CAAAAL19524-06
3-Bromo-4-nitropyridine-N-oxide 98+%
3-Bromo-4-nitropyridine N-oxide
Formula: C₅H₃BrN₂O₃
MW: 219.00 g/mol
Melting Pt: 152…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00085925
CAS Number: 1678-49-5
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR