(S)-1-Benzyl-3-pyrrolidinol 99%, ee 99%

Warning

Synonyms: (S)-1-Benzyl-3-hydroxypyrrolidine(S)-1-Benzylpyrrolidin-3-ol

L19497-06 L19497-03 L19497-03 L19497-06
AAAL19497-03EA 39.24 USD
CAAAAL19497-03 AAAL19497-03 CAAAAL19497-06 AAAL19497-06
(S)-1-Benzyl-3-pyrrolidinol 99%, ee 99%
(S)-1-Benzyl-3-pyrrolidinol

For synthesis of optically active products

Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
Boiling Pt: 116 °C/0.9 mm
Density: 1.070
Flash Pt: 148 °C(298 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075485
CAS Number: 101385-90-4
EINECS: 212-273-5

Order Now

Specification Test Results

Optical rotation -3.7° (c=5 in methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR