Boc-cycloleucine 98%

Synonyms: 1-N-BOC-Aminocyclopentanecarboxylic acid1-(Boc-amino)cyclopentanecarboxylic acid1-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

L19483-03 L19483-06 L19483-06 L19483-03
AAAL19483-03EA 44.29 USD
CAAAAL19483-03 CAAAAL19483-06 AAAL19483-03 AAAL19483-06
Boc-cycloleucine 98%
Boc-cycloleucine
Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
Melting Pt: 131…134 °C
Storage Temperature: Ambient
MDL Number: MFCD01076126
CAS Number: 35264-09-6
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR