(R)-(+)-N-Benzyl-α-methylbenzylamine ≥99%, ee 96%, ChiPros®
|
Danger
|
Synonyms:
(R)-(+)-N-Benzyl-1-phenylethylamine
, (R)-N-Benzyl-1-phenylethanamine
L19298-06
L19298-14
CAAAAL19298-06
CAAAAL19298-14
(R)-(+)-N-Benzyl-α-methylbenzylamine ≥99%, ee 96%, ChiPros®
(R)-(+)-N-Benzyl-α-methylbenzylamine
Formula:
C₁₅H₁₇N MW: 211.31 g/mol Boiling Pt: 171 °C/15 mm Density: 1.010 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015010 CAS Number: 38235-77-7 EINECS: 609-537-0 UN: 2735 ADR: "8",III |
Specification Test Results
Optical rotation | +39° (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...