5-Methoxy-1-methyl-1H-indole-3-carbaldehyde 98%

Warning

Synonyms: 5-Methoxy-1-methylindole-3-carboxaldehyde

L19269-03 L19269-MD L19269-MD L19269-03
AAAL19269-MDEA 61.22 USD
CAAAAL19269-MD AAAL19269-MD CAAAAL19269-03 AAAL19269-03
5-Methoxy-1-methyl-1H-indole-3-carbaldehyde 98%
5-Methoxy-1-methyl-1H-indole-3-carbaldehyde
Formula: C₁₁H₁₁NO₂
MW: 189.214 g/mol
Melting Pt: 133…136 °C
Storage Temperature: Ambient
MDL Number: MFCD01578324
CAS Number: 39974-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR