3-Fluoro-4-methylbenzylamine 95%

Danger

L19171-MD L19171-03 L19171-MD L19171-03
AAAL19171-03EA 100.67 USD
CAAAAL19171-MD AAAL19171-03 AAAL19171-MD CAAAAL19171-03
3-Fluoro-4-methylbenzylamine 95%
3-Fluoro-4-methylbenzylamine
Formula: C₈H₁₀FN
Density: 1.050
Storage Temperature: Ambient
MDL Number: MFCD01631531
CAS Number: 261951-67-1
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR