4-(Difluoromethoxy)benzylamine 97%

Danger

Synonyms: (4-(Difluoromethoxy)phenyl)methanamine

L19166-03 L19166-MD L19166-06 L19166-06 L19166-MD L19166-03
AAAL19166-MDEA 48.47 USD
AAAL19166-MD AAAL19166-06 CAAAAL19166-MD CAAAAL19166-06 AAAL19166-03 CAAAAL19166-03
4-(Difluoromethoxy)benzylamine 97%
4-(Difluoromethoxy)benzylamine
Formula: C₈H₉F₂NO
MW: 173.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236231
CAS Number: 177842-14-7
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR