2-(Difluoromethoxy)benzylamine 97%

Supplier: Thermo Scientific Chemicals

Danger

Synonyms: (2-(Difluoromethoxy)phenyl)methanamine

L19165-03 L19165-MD L19165-03 L19165-MD
AAAL19165-MDEA 44.33 USD
AAAL19165-MD CAAAAL19165-MD AAAL19165-03 CAAAAL19165-03
2-(Difluoromethoxy)benzylamine 97%
2-(Difluoromethoxy)benzylamine
Formula: C₈H₉F₂NO
MW: 173.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236229
CAS Number: 243863-36-7
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR