1H-Pyrrole-2-carbaldehyde oxime 97%

L18959-14 L18959-03 L18959-03 L18959-06 L18959-14 L18959-06
AAAL18959-14EA 271.95 USD
CAAAAL18959-03 CAAAAL18959-14 CAAAAL18959-06 AAAL18959-14 AAAL18959-03 AAAL18959-06
1H-Pyrrole-2-carbaldehyde oxime 97%
1H-Pyrrole-2-carbaldehyde oxime
Formula: C₅H₆N₂O
MW: 110.12 g/mol
Melting Pt: 163…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00965095
CAS Number: 32597-34-5
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR