4-Benzyloxy-3-fluorophenylboronic acid 98%

Warning

Synonyms: 4-Benzyloxy-3-fluorobenzeneboronic acid

L18520-MD L18520-MD L18520-03 L18520-03
AAAL18520-MDEA 46 USD
CAAAAL18520-MD AAAL18520-MD CAAAAL18520-03 AAAL18520-03
4-Benzyloxy-3-fluorophenylboronic acid 98%
4-Benzyloxy-3-fluorophenylboronic acid
Formula: C₁₃H₁₂BFO₃
MW: 246.04 g/mol
Melting Pt: 129…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00994627
CAS Number: 133057-83-7
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR