4-Pentylphenylboronic acid 97%

Warning

Synonyms: 4-n-Pentylbenzeneboronic acid(4-Pentylphenyl)boronic acid

L18011-03 L18011-06 L18011-MD L18011-03 L18011-06 L18011-MD
AAAL18011-03EA 165.39 USD
CAAAAL18011-MD AAAL18011-03 CAAAAL18011-03 AAAL18011-MD AAAL18011-06 CAAAAL18011-06
4-Pentylphenylboronic acid 97%
4-Pentylphenylboronic acid
Formula: C₁₁H₁₇BO₂
MW: 192.07 g/mol
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00995151
CAS Number: 121219-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR