1-(Trifluoromethyl)cyclopropanecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-(Trifluoromethyl)cyclopropane-1-carboxylic acid

L17957-03 L17957-MD L17957-03 L17957-MD
AAAL17957-MDEA 106.52 USD
CAAAAL17957-MD AAAL17957-MD CAAAAL17957-03 AAAL17957-03
1-(Trifluoromethyl)cyclopropanecarboxylic acid 97%
1-(Trifluoromethyl)cyclopropanecarboxylic acid
Formula: C₅H₅F₃O₂
MW: 154.09 g/mol
Boiling Pt: 182…184 °C
Melting Pt: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD03093070
CAS Number: 277756-46-4
EINECS: 000-000-0
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR