3-Fluoro-4-formylphenylboronic acid 98%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 3-Fluoro-4-formylbenzeneboronic acid

L17851-03 L17851-03
AAAL17851-03EA 40.7 USD
AAAL17851-03 CAAAAL17851-03
3-Fluoro-4-formylphenylboronic acid 98%
3-Fluoro-4-formylphenylboronic acid
Formula: C₇H₆BFO₃
MW: 167.93 g/mol
Melting Pt: ca 240 °C
Storage Temperature: Ambient
MDL Number: MFCD02093051
CAS Number: 248270-25-9
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR