Methyl-4-fluorosalicylate 98+%

Warning

Synonyms: Methyl 4-fluoro-2-hydroxybenzoate

L17846-03 L17846-03 L17846-MD L17846-MD
AAAL17846-MDEA 68.73 USD
CAAAAL17846-MD AAAL17846-MD CAAAAL17846-03 AAAL17846-03
Methyl-4-fluorosalicylate 98+%
Methyl 4-fluorosalicylate
Formula: C₈H₇FO₃
MW: 170.14 g/mol
Melting Pt: 38…40 °C
Storage Temperature: Ambient
MDL Number: MFCD02093965
CAS Number: 392-04-1
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR