4-Fluoro-3-formylphenylboronic acid 98+%

Warning

Synonyms: (4-Fluoro-3-formylphenyl)boronic acid4-Fluoro-3-formylbenzeneboronic acid

L17808-06 L17808-03 L17808-06
AAL17808-06EA 294.79 USD
AAL17808-06 AAL17808-03 CAAAL17808-06
4-Fluoro-3-formylphenylboronic acid 98+%
4-Fluoro-3-formylphenylboronic acid
Formula: C₇H₆BFO₃
MW: 167.93 g/mol
Melting Pt: ca 240 °C
Storage Temperature: Ambient
MDL Number: MFCD02093074
CAS Number: 374538-01-9
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR