Ethyl-3-(benzylamino)propionate 95%

Warning

Synonyms: N-Benzyl-beta-alanine Ethyl EsterN-Benzyl-3-aminopropionic Acid Ethyl EsterEthyl 3-(benzylamino)propanoate

L17652-06 L17652-14 L17652-14 L17652-06 L17652-03 L17652-03
AAAL17652-14EA 101.52 USD
CAAAAL17652-06 AAAL17652-14 AAAL17652-03 CAAAAL17652-14 CAAAAL17652-03 AAAL17652-06
Ethyl-3-(benzylamino)propionate 95%
Ethyl-3-(benzylamino)propionate
Formula: C₁₂H₁₇NO₂
MW: 207.27 g/mol
Boiling Pt: 150 °C/0.2 mm
Density: 1.030
Storage Temperature: Ambient
MDL Number: MFCD00085541
CAS Number: 23583-21-3
EINECS: 245-759-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR