6-Methyl-1-indanone 98%

L17559-03 L17559-06 L17559-03 L17559-06
AAAL17559-03EA 55.61 USD
AAAL17559-03 AAAL17559-06 CAAAAL17559-06 CAAAAL17559-03
6-Methyl-1-indanone 98%
6-Methyl-1-indanone
Formula: C₁₀H₁₀O
MW: 146.19 g/mol
Boiling Pt: 124…126 °C/11 mm
Melting Pt: 57…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00275729
CAS Number: 24623-20-9
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR