Ethyl-5-chloro-1H-indole-2-carboxylate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 5-chloro-2-indolecarboxylateEthyl 5-chloro-1H-indole-2-carboxylate

L17554-03 L17554-03 L17554-06 L17554-14 L17554-06
AAAL17554-06EA 119.62 USD
AAAL17554-06 CAAAAL17554-06 AAAL17554-03 CAAAAL17554-03 CAAAAL17554-14
Ethyl-5-chloro-1H-indole-2-carboxylate 98%
Ethyl-5-chloro-1H-indole-2-carboxylate
Formula: C₁₁H₁₀ClNO₂
MW: 223.66 g/mol
Melting Pt: 166…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00005610
CAS Number: 4792-67-0
EINECS: 225-345-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR