4-Isopropylphenylboronic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Isopropylbenzeneboronic acid

L17459-03 L17459-06 L17459-06 L17459-03
AAAL17459-06EA 427.99 USD
CAAAAL17459-03 AAAL17459-06 AAAL17459-03 CAAAAL17459-06
4-Isopropylphenylboronic acid 98+%
4-Isopropylphenylboronic acid
Formula: C₉H₁₃BO₂
MW: 164.01 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD01074614
CAS Number: 16152-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR