2,4,6-Trimethylbenzeneboronic acid neopentyl glycol ester 99%

L17232-06 L17232-03 L17232-06 L17232-03
AAAL17232-06EA 116.55 USD
CAAAAL17232-03 AAAL17232-06 AAAL17232-03 CAAAAL17232-06
2,4,6-Trimethylbenzeneboronic acid neopentyl glycol ester 99%
2,4,6-Trimethylbenzeneboronic acid neopentyl glycol ester
Formula: C₁₄H₂₁BO₂
MW: 232.13 g/mol
Melting Pt: 46…48 °C
Storage Temperature: Ambient
MDL Number: MFCD01318178
CAS Number: 214360-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR