1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene 96%

Warning

Synonyms: 2,2'-(Benzene-1,3-diyl)bis(1,1,1,3,3,3-hexafluoropropan-2-ol)

L16894-06 L16894-06 L16894-14 L16894-14
AAAL16894-14EA 208.06 USD
CAAAAL16894-14 AAAL16894-14 CAAAAL16894-06 AAAL16894-06
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene 96%
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene
Formula: C₁₂H₆F₁₂O₂
MW: 410.16
Boiling Pt: 99…100 °C/20 mm
Melting Pt: 9…10 °C
Density: 1.659
Flash Pt: 97 °C(206 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042090
CAS Number: 802-93-7
EINECS: 212-354-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR