Chlorodimethylpropylsilane 97%

Danger

Synonyms: PropyldimethylchlorosilaneDimethylpropylchlorosilane

L16621-14 L16621-14 L16621-06 L16621-06
AAAL16621-14EA 93.01 USD
AAAL16621-14 CAAAAL16621-14 AAAL16621-06 CAAAAL16621-06
Chlorodimethylpropylsilane 97%
Chlorodimethylpropylsilane
Formula: C₅H₁₃ClSi
MW: 136.69 g/mol
Boiling Pt: 113…114 °C
Density: 0.873
Flash Pt: 8 °C(46 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039301
CAS Number: 17477-29-1
EINECS: 241-492-9
UN: 2985
ADR: "3",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR