Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethylethoxysilane 94% ammonia free

Supplier: Alfa Aesar
Danger

L16476-22 L16476-22 L16476-14 L16476-14
AAAL16476-22EA 358.44 USD InStock
CAAAAL16476-22 AAAL16476-22 CAAAAL16476-14 AAAL16476-14
Dimethylethoxysilane 94% ammonia free
Dimethylethoxysilane


Formula: C₄H₁₂OSi
MW: 104,23
Boiling Pt: 55°C
Density: 0.757
Flash Pt: 15°C (59°F)
Storage Temperature: Ambient
MDL Number: MFCD00026750
CAS Number: 14857-34-2
EINECS: 238-921-7
UN: 1993
ADR: 3,II
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty