3-(2-Carboxyvinyl)phenylboronic acid 97%

Warning

Synonyms: 3-(3-Boronophenyl)acrylic acid3-(2-Carboxyvinyl)benzeneboronic acid

L16369-MD L16369-MD L16369-03 L16369-03
AAAL16369-MDEA 76.23 USD
CAAAAL16369-03 CAAAAL16369-MD AAAL16369-MD AAAL16369-03
3-(2-Carboxyvinyl)phenylboronic acid 97%
3-(2-Carboxyvinyl)phenylboronic acid
Formula: C₉H₉BO₄
MW: 191.98 g/mol
Melting Pt: ca 218 °C
Storage Temperature: Ambient
MDL Number: MFCD01075738
CAS Number: 216144-91-1
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR