3-(4-Fluorophenyl)-1-isopropylindole 98%

Synonyms: 3-(4-Fluorophenyl)-1-isopropyl-1H-indole

L16341-03 L16341-06 L16341-06 L16341-03
AAAL16341-03EA 27.36 USD
CAAAAL16341-03 AAAL16341-03 CAAAAL16341-06 AAAL16341-06
3-(4-Fluorophenyl)-1-isopropylindole 98%
3-(4-Fluorophenyl)-1-isopropylindole
Formula: C₁₇H₁₆FN
MW: 253.32 g/mol
Melting Pt: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD01075733
CAS Number: 93957-49-4
EINECS: 418-790-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR