N-(4-Bromobutoxy)phthalimide ≥98%

Warning

L16314-06 L16314-03 L16314-06 L16314-03
AAAL16314-06EA 395.22 USD
AAAL16314-06 CAAAAL16314-06 AAAL16314-03 CAAAAL16314-03
N-(4-Bromobutoxy)phthalimide ≥98%
N-(4-Bromobutoxy)phthalimide
Formula: C₁₂H₁₂BrNO₃
MW: 298.14 g/mol
Melting Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00517028
CAS Number: 5093-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR