3-Methyl-4-isoxazolecarboxylic acid 98+%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 3-Methylisoxazole-4-carboxylic acid

L16307-MD L16307-03 L16307-MD L16307-03
AAAL16307-03EA 170 USD
AAAL16307-03 CAAAAL16307-03 AAAL16307-MD CAAAAL16307-MD
3-Methyl-4-isoxazolecarboxylic acid 98+%
3-Methyl-4-isoxazolecarboxylic acid
Formula: C₅H₅NO₃
MW: 127.10 g/mol
Melting Pt: 185…187 °C
Storage Temperature: Ambient
MDL Number: MFCD01318161
CAS Number: 17153-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR