N-Boc-D-prolinol 99%
|
Warning
|
Synonyms:
(R)-(+)-1-Boc-2-pyrrolidinemethanol
, (R)-1-Boc-2-pyrrolidinemethanol
, N-t-Boc-D-prolinol
, (R)-(+)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol
, N-tert-Butoxycarbonyl-D-prolinol
, Boc-D-Prolinol
, (R)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol
L16268-03
CAAAAL16268-03
N-Boc-D-prolinol 99%
N-Boc-D-prolinol
Formula:
C₁₀H₁₉NO₃ MW: 201.3 g/mol Melting Pt: 59…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040580 CAS Number: 83435-58-9 |
Specification Test Results
Optical rotation | +54° (c=5 in methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...