3-Phenylphthalide 99%

Synonyms: 3-Phenylisobenzofuran-1(3H)-one

L16181-14 L16181-06 L16181-06 L16181-14
AAAL16181-14EA 199.68 USD
CAAAAL16181-06 AAAL16181-14 CAAAAL16181-14 AAAL16181-06
3-Phenylphthalide 99%
3-Phenylphthalide
Formula: C₁₄H₁₀O₂
MW: 210.23
Melting Pt: 116…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00023099
CAS Number: 5398-11-8
EINECS: 226-426-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR