1-Vinylimidazole 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Vinyl-1H-imidazole
L16174-14
L16174-14
L16174-22
L16174-22
AAAL16174-22EA
77.5
USD
AAAL16174-22
CAAAAL16174-22
CAAAAL16174-14
AAAL16174-14
1-Vinylimidazole 99%
1-Vinylimidazole
Formula:
C₅H₆N₂ MW: 94.12 g/mol Boiling Pt: 192…194 °C Melting Pt: < –50 °C Density: 1.038 Flash Pt: 84 °C(183 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005297 CAS Number: 1072-63-5 EINECS: 214-012-0 UN: 3267 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...