Methyl pentafluorophenyl carbonate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Carbonic Acid Methyl Pentafluorophenyl EsterMethyl Pentafluorophenyl Carbonate

L16107-14 L16107-06 L16107-06 L16107-14
AAAL16107-14EA 312.55 USD
CAAAAL16107-06 AAAL16107-14 AAAL16107-06 CAAAAL16107-14
Methyl pentafluorophenyl carbonate 97%
Methyl pentafluorophenyl carbonate
Formula: C₈H₃F₅O₃
MW: 242.10 g/mol
Boiling Pt: 84 °C/3 mm
Melting Pt: 27…31 °C
Storage Temperature: Ambient
MDL Number: MFCD01075723
CAS Number: 36919-03-6
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR