4,4'-Bipiperidine dihydrochloride 99%

Warning

L15696-14 L15696-14 L15696-06 L15696-06
AAAL15696-14EA 185.03 USD
CAAAAL15696-14 CAAAAL15696-06 AAAL15696-14 AAAL15696-06
4,4'-Bipiperidine dihydrochloride 99%
4,4'-Bipiperidine dihydrochloride
Formula: C₁₀H₂₀N₂·2HCl
MW: 241.21 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00005998
CAS Number: 78619-84-8
EINECS: 278-958-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR