2-Fluoro-4-biphenylboronic acid 97%
|
Warning
|
Synonyms:
2-Fluoro-1,1'-biphenyl-4-ylboronic acid
, (3-fluoro-4-phenyl)phenylboronic acid
, 3-Fluoro-4-phenylbenzeneboronic acid
, 2-Fluoro-4-biphenylylboronic acid
L15634-03
L15634-03
L15634-06
L15634-06
AAAL15634-06EA
157.52
USD
CAAAAL15634-03
AAAL15634-06
AAAL15634-03
CAAAAL15634-06
2-Fluoro-4-biphenylboronic acid 97%
2-Fluoro-4-biphenylboronic acid
Formula:
C₁₂H₁₀BFO₂ MW: 216.02 g/mol Melting Pt: ca 242…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075707 CAS Number: 178305-99-2 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...