4-(2-Carboxyvinyl)phenylboronic acid 95%

Warning

Synonyms: 4-(2-Carboxyvinyl)benzeneboronic acid3-(4-Boronophenyl)acrylic acid

L15586-MD L15586-03 L15586-06 L15586-MD L15586-03 L15586-06
AAAL15586-MDEA 60.88 USD
AAAL15586-MD CAAAAL15586-MD AAAL15586-06 AAAL15586-03 CAAAAL15586-06 CAAAAL15586-03
4-(2-Carboxyvinyl)phenylboronic acid 95%
4-(2-Carboxyvinyl)phenylboronic acid
Formula: C₉H₉BO₄
MW: 191.98 g/mol
Melting Pt: ca 190…196 °C
Storage Temperature: Ambient
MDL Number: MFCD01075704
CAS Number: 151169-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR