6,6-Dimethyl-5,7-dioxaspiro[2.5]octane-4,8-dione 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Cyclopropane-1,1-dicarboxylic Acid cycl-Isopropylidene Ester
, cycl-Isopropylidene Cyclopropane-1,1-dicarboxylate
L15571-03
L15571-03
L15571-06
L15571-06
AAAL15571-03EA
31.54
USD
AAAL15571-03
AAAL15571-06
CAAAAL15571-06
CAAAAL15571-03
6,6-Dimethyl-5,7-dioxaspiro[2.5]octane-4,8-dione 99%
6,6-Dimethyl-5,7-dioxaspiro[2.5]octane-4,8-dione
Formula:
C₈H₁₀O₄ MW: 170.16 g/mol Melting Pt: 63…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042796 CAS Number: 5617-70-9 EINECS: 227-044-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...