Thiophene-3-carboxaldehyde diethyl acetal 98%

L15520-03 L15520-03 L15520-06 L15520-06
AAAL15520-06EA 118.77 USD
AAAL15520-06 AAAL15520-03 CAAAAL15520-06 CAAAAL15520-03
Thiophene-3-carboxaldehyde diethyl acetal 98%
Thiophene-3-carboxaldehyde diethyl acetal
Formula: C₉H₁₄O₂S
MW: 186.27 g/mol
Boiling Pt: 130 °C/30 mm
Storage Temperature: Ambient
MDL Number: MFCD01318173
CAS Number: 3199-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR