Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9-BBN dimer ≥98%

Supplier: Alfa Aesar
Danger

L15166-14 L15166-14 L15166-06 L15166-06
AAAL15166-14EA 374.92 USD InStock
AAAL15166-14 CAAAAL15166-14 CAAAAL15166-06 AAAL15166-06
9-BBN dimer ≥98%
9-BBN dimer


Formula: C₁₆H₃₀B₂
MW: 244,04
Melting Pt: 149…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00168069
CAS Number: 21205-91-4
UN: 3393
ADR: 4.2,I
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty