Rhodium(II) acetate dimer 98+%
|
Warning
|
Synonyms:
Tetrakis(acetato)dirhodium(II)
, Dirhodium tetraacetate
L15152-ME
L15152-MC
L15152-MC
L15152-ME
AAAL15152-MEEA
577.99
USD
AAAL15152-ME
AAAL15152-MC
CAAAAL15152-ME
CAAAAL15152-MC
Rhodium(II) acetate dimer 98+%
Rhodium (II) acetate dimer
Formula:
C₈H₁₂O₈Rh₂ MW: 441.987 g/mol Melting Pt: 205 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003538 CAS Number: 15956-28-2 EINECS: 240-084-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...