Lithiumbis(trimethylsilyl)amide 20% ∼1.06 M in THF/ethylbenzene

Supplier: Thermo Scientific Chemicals
Danger

L15012-AE L15012-AE
AAAL15012-AEEA 107.99 USD
CAAAAL15012-AE AAAL15012-AE
Lithiumbis(trimethylsilyl)amide 20% ∼1.06 M in THF/ethylbenzene
Lithium bis(trimethylsilyl)amide

Packaged in resealable septum cap bottle

Formula: C₆H₁₈LiNSi₂
MW: 167.33 g/mol
Density: 0.891
Flash Pt: -17 °C(1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008261
CAS Number: 4039-32-1
EINECS: 223-725-6
UN: 2924
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR