2,4-Dihydroxy-3-nitropyridine 99%

Warning

Synonyms: 3-Nitropyridine-2,4-diol

L14933-06 L14933-14 L14933-06 L14933-03 L14933-14 L14933-03
AAAL14933-06EA 102.61 USD
AAAL14933-06 CAAAAL14933-14 CAAAAL14933-03 CAAAAL14933-06 AAAL14933-14 AAAL14933-03
2,4-Dihydroxy-3-nitropyridine 99%
2,4-Dihydroxy-3-nitropyridine
Formula: C₅H₄N₂O₄
MW: 156.10 g/mol
Melting Pt: ca 265 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01075671
CAS Number: 89282-12-2
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR