Cyclopropanecarboxaldehyde 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cyclopropanecarbaldehyde

L14931-06 L14931-03
AAAL14931-06EA 124.99 USD
AAAL14931-06 AAAL14931-03
Cyclopropanecarboxaldehyde 98%
Cyclopropanecarboxaldehyde
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 98…101 °C
Density: 0.938
Flash Pt: 7 °C(44 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012261
CAS Number: 1489-69-6
EINECS: 000-000-0
UN: 2924
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR