2-Methoxy-5-isopropylbenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Isopropyl-2-methoxybenzaldehyde

L14737-14 L14737-06 L14737-14 L14737-22 L14737-22 L14737-06
AAAL14737-14EA 154.99 USD
AAAL14737-14 CAAAAL14737-06 AAAL14737-22 CAAAAL14737-14 AAAL14737-06 CAAAAL14737-22
2-Methoxy-5-isopropylbenzaldehyde 97%
2-Methoxy-5-isopropylbenzaldehyde
Formula: C₁₁H₁₄O₂
MW: 178.23
Boiling Pt: 96 °C/1 mm
Storage Temperature: Ambient
MDL Number: MFCD03095324
CAS Number: 85902-68-7
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR