Ethyl-2-(3-N-phthalimidopropyl)acetoacetate 95%

L14724-14 L14724-06 L14724-06 L14724-14
AAAL14724-14EA 192.36 USD
AAAL14724-14 CAAAAL14724-06 CAAAAL14724-14 AAAL14724-06
Ethyl-2-(3-N-phthalimidopropyl)acetoacetate 95%
Ethyl-2-(3-N-phthalimidopropyl)acetoacetate
Formula: C₁₇H₁₉NO₅
MW: 317.34 g/mol
Melting Pt: 62…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00399474
CAS Number: 55747-45-0
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR