Dibenzyl-5-aminoisophthalate 96%

L14629-06 L14629-06 L14629-03 L14629-03
AAAL14629-03EA 46.34 USD
AAAL14629-03 CAAAAL14629-06 AAAL14629-06 CAAAAL14629-03
Dibenzyl-5-aminoisophthalate 96%
Dibenzyl-5-aminoisophthalate
Formula: C₂₂H₁₉NO₄
MW: 361.40 g/mol
Melting Pt: 116…118 °C
Storage Temperature: Ambient
MDL Number: MFCD03093065
CAS Number: 152699-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR