2-Amino-6-methyl-4(3H)-quinazolone 96%

Supplier: Thermo Scientific Chemicals
Warning

L14558-MD L14558-03 L14558-03 L14558-MD
AAAL14558-03EA 138.99 USD
AAAL14558-03 CAAAAL14558-03 AAAL14558-MD CAAAAL14558-MD
2-Amino-6-methyl-4(3H)-quinazolone 96%
2-Amino-6-methyl-4(3H)-quinazolone
Formula: C₉H₉N₃O
MW: 175.19 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00236015
CAS Number: 50440-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR